missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14985-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.Specifications
PPA1 | |
Polyclonal | |
Unconjugated | |
PPA1 | |
Inorganic pyrophosphatase, IOPPP, PP, PP1, PPA1, PPase, pyrophosphatase (inorganic) 1, SID6 8061 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294504, 5464, 67895 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15181, Q9D819 | |
PPA1 | |
PPA1 Fusion Protein Ag6969 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |