missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20635-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PPAPDC3, also known as nuclear envelope transmembrane protein 39 (NET39), was initially discovered in an in silico screen for secreted or membrane proteins. It is a member of the PAP2 superfamily of phosphatases and haloperoxidases. PPAPDC3 has recently been shown to act as a negative regulator of myoblast differentiation by diminishing the activity of the mammalian target of rapamycin TOR. PPAPDC3 is highly expressed in cardiac and skeletal muscle and becomes strongly upregulated during cultured myoblast differentiation tissues. Overexpression of PPAPDC3 in myoblasts repressed myogenesis while knockdown by RNA interference promoted differentiation indicating its part in the regulatory mechanism for myogenesis.Specifications
PPAPDC3 | |
Polyclonal | |
Unconjugated | |
PLPP7 | |
C9orf67, KIAA0515, NET39, PPAPDC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227721, 296635, 84814 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5FVJ3, Q8NBV4, Q91WB2 | |
Plpp7 | |
PPAPDC3 Fusion Protein Ag14676 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |