missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPARA (Human) Recombinant Protein
Human PPARA full-length ORF ( AAH00052, 1 a.a. - 258 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005465-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVDTESPLCPLSPLEAGDLESPLSEEFLQEMGNIQEISQSIGEDSSGSFGFTEYQYLGSCPGSDGSVITDTLSPASSPSSVTYPVVPGSVDESPSGALNIECRICGDKASGYHYGVHACEGCKGFFRRTIRLKLVYDKCDRSCKIQKKNRNKCQYCRFHKCLSVGRSHNAIRFGRMPRSEKAKLKAEILTCEHDIEDSETADLKSLAKRIYEAYLKNFNMNKVKARVILSGKASNNPVGVCGCSGFSWQHGTSVVEDD
Specifications
AAH00052 | |
peroxisome proliferator-activated receptor alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPARA | |
GST |
5465 | |
Wheat germ expression system | |
10 μg | |
MGC2237, MGC2452, NR1C1, PPAR, hPPAR | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |