Läs mer
Tocris Bioscience™ PPDA
GREENER_CHOICE
Produktkod. 18085752
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18085752 50 mg
18054802 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18085752 Leverantör Tocris Bioscience™ Leverantörsnummer 2530/50
GluN2C/GluN2D-selective NMDA antagonist
Subtype-selective NMDA receptor antagonist that preferentially binds to GluN2C/GluN2D (formally NR2C/NR2D) containing receptors (Ki values are 0.096, 0.125, 0.31 and 0.55 μM for GluN2C, GluN2D, GluN2B and GluN2A subunits respectively).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 684283-16-7
Molekylformel C21H18N2O5
Specifikationer
Kemiskt namn eller material PPDA
Kvantitet 50 mg
Formel vikt 378.38
Renhet >98%
Molekylvikt (g/mol) 378.384