Learn More
PPEF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12875-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850824
  • 1656.73 SEK / 20µL
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
The PPEF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPEF1. This antibody recognizes human, mouse antigen. The PPEF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PPEF1
Polyclonal
Unconjugated
20 μL
5475
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL
Rabbit
Primary
Human, Mouse