Learn More
PPEF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12875-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The PPEF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPEF1. This antibody recognizes human, mouse antigen. The PPEF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PPEF1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
5475 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
267 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |