Learn More
PPP1CC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11082-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
DYRK1B is a member of the DYRK family of protein kinases. DYRK1B contains a bipartite nuclear localization signal and is found mainly in muscle and testis. The protein is proposed to be involved in the regulation of nuclear functions. Three isoforms of DYRK1B have been identified differing in the presence of two alternatively spliced exons within the catalytic domain.Specifications
PPP1CC | |
Polyclonal | |
Unconjugated | |
PPP1CC | |
PP1 Gamma, PP 1G, PPP1CC, PPP1G | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19047, 24669, 5501 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36873, P63087 | |
Ppp1cc | |
PPP1CC Fusion Protein Ag1550 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |