missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R42 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555743
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LTHLYLQNNC ISCIENLRSL KKLEKLYLGG NYIAVIEGLE GLGELRELHV ENQRLPLGEK LLFDPRTL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
Specifications
PPP1R42 | |
Polyclonal | |
Unconjugated | |
PPP1R42 | |
PPP1R42 | |
Recombit protein corresponding to Human PPP1R42 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z4L9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286187 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only