missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ PPP1R7 Monoclonal Antibody (OTI4F9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524968
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PPP1R7 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase 1 (PP1) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit, and a catalytic subunit. Sds22, also known as PPP1R7 (protein phosphatase 1, regulatory (inhibitor) subunit 7), is a 360 amino acid protein that localizes to the nucleus and contains ten LRR (leucine rich) repeats. Expressed in a variety of tissues, Sds22 functions as a regulatory subunit of the PP1 complex, suggesting a role in protein regulation throughout the cell. Multiple isoforms of Sds22 exist due to alternative splicing events.
Specifikationer
| PPP1R7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15435, Q5HZV9 | |
| PPP1R7 | |
| Full length human recombinant protein of PPP1R7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4F9 | |
| Unconjugated | |
| PPP1R7 | |
| 2310014J01Rik; hypothetical protein LOC560323; im:7136431; Ppp1r7; Protein phosphatase 1 regulatory subunit 22; protein phosphatase 1 regulatory subunit 7; protein phosphatase 1, regulatory (inhibitor) subunit 7; protein phosphatase 1, regulatory subunit 7; protein phosphatase-1 regulatory subunit 7; Sds22; testis secretory sperm-binding protein Li 210a; zgc:123325 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100855698, 301618, 5510 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering