missing translation for 'onlineSavingsMsg'
Learn More

PPP2R2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16569-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826224

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
Specifications

Specifications

PPP2R2A
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36876, P63151, Q6P1F6
B55 ALPHA, B55A, PP2A subunit B isoform alpha, PPP2R2A, PR52A, PR55A
Rabbit
IgG
150 μL
-20°C
Primary
117104, 5520, 71978
Immunoprecipitation, Western Blot
Unconjugated
PPP2R2A
Liquid
PPP2R2A
PPP2R2A Fusion Protein Ag9851
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers