Learn More
PPP3CC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55163-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PPP3CC.
Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca, calcineurin A-beta, and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).Specifications
PPP3CC | |
Polyclonal | |
Unconjugated | |
PPP3CC | |
36 53aa, CALNA3, CAM PRP catalytic subunit, CNA3, PPP3CC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19057, 5533 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48454 | |
Ppp3cc | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |