Learn More
PPP4R1 Mouse anti-Human, Unconjugated, Clone: 3D12G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67392-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839995
  • 1236.20 SEK / 20µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The PPP4R1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPP4R1. This antibody recognizes human, mouse, rat antigen. The PPP4R1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PPP4R1
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
3D12G12
Unconjugated
20 μL
9989
IgG1