Learn More
PPP4R1 Mouse anti-Human, Unconjugated, Clone: 3D12G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67392-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The PPP4R1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPP4R1. This antibody recognizes human, mouse, rat antigen. The PPP4R1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PPP4R1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
3D12G12 | |
Unconjugated | |
150 μL | |
9989 | |
IgG1 |