missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15852-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Distinct DNA repair pathways minimize the consequences of mutagenic events. Reactive oxygen species (ROS) are highly reactive atoms with an unpaired electron that are conducive to double-strand DNA breaking events. Artemis, named after the Greek goddess for the protection of children, is one of the major proteins contributing to the preservation of double-strand breaks in DNA by cutting away the damaged parts of the DNA, which allows the strands to rejoin. Artemis is a single-strand-specific 5′ to 3′ exonuclease that forms a complex with the 469 kDa DNA-dependent protein kinase (DNA-PKcs). DNA-PKcs phosphorylates Artemis, and Artemis acquires endonucleolytic activity on 5′ and 3′ overhangs and hairpins. These activities are essential for V(D)J recombination and for the 5′ and 3′ overhang processing in nonhomologous DNA end joining.Specifications
PPP6C | |
Polyclonal | |
Unconjugated | |
PPP6C | |
PP6C, PPP6, PPP6C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171121, 5537, 67857 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00743, Q64620, Q9CQR6 | |
Ppp6c | |
PPP6C Fusion Protein Ag8670 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |