Läs mer
Tocris Bioscience™ PPY A
GREENER_CHOICE
Produktkod. 30001684
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001684 10 mg
30001521 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001684 Leverantör Tocris Bioscience™ Leverantörsnummer 4730/10
 Begränsad artikel
Beräknad leverans: 31-08-2026
Lägg i varukorgen
Lägg i varukorgen
Potent inhibitor of Abl T315l mutant and wild-type Abl kinases
Potent inhibitor of T315l mutant and wild-type Abl kinases (IC50 values are 9 and 20 nM, respectively). Inhibits growth of cells transformed with either the Bcr-Abl T315l mutant or wild-type Bcr-Abl gene.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 875634-01-8
Molekylformel C22H20N4O2
Specifikationer
Kemiskt namn eller material PPY A
Kvantitet 10 mg
Formel vikt 372.42
Renhet >98%
Molekylvikt (g/mol) 372.428