Läs mer
Tocris Bioscience™ PR 619
Broad spectrum DUB inhibitor
Brand: Tocris Bioscience™ 4482/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.Specifikationer
| PR 619 | |
| 2,6-Diamino-3,5-dithiocyanopyridine | |
| Soluble to 10mM in DMSO | |
| Bioactive Small Molecule |
| 2645-32-1 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use