Läs mer
Praeruptorin A, TRC
Produktkod. 30010958
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010958 100 mg
30011609 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010958 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP699445100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73069-25-7
Molekylformel C21 H22 O7
Molekylvikt (g/mol) 386.4
Synonym 2-Butenoic acid, 2-methyl-, (9R,10R)-10-(acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-9-yl ester, (2Z)-rel-, 2-Butenoic acid, 2-methyl-, 10-(acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-9-yl ester, [9α(Z),10α]-(±)-, 2H,8H-Benzo[1,2-b:3,4-b']dipyran, 2-butenoic acid deriv., (±)-Praeruptorin A, 2-Butenoic acid, 2-methyl-, 10-(acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-9-yl ester, [9α(Z),10α]-, dl-Praeruptorin A, Praeruptorin A, 2-Methyl-2-butenoic Acid [9α(Z),10α]-(±)-10-(Acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-9-yl Ester, 2H,8H-Benzo[1,2-b:3,4-b']dipyran-2-butenoic Acid deriv., 2-Methyl-2-Butenoic Acid [9α(Z),10α]-10-(Acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-9-yl Ester;
IUPAC-namn [(9S,10S)-10-acetoxy-8,8-dimethyl-2-oxo-9,10-dihydropyrano[2,3-f]chromen-9-yl] (Z)-2-methylbut-2-enoate
LEDER C\C=C(\C)/C(=O)O[C@H]1[C@@H](OC(=O)C)c2c(OC1(C)C)ccc3C=CC(=O)Oc23
Specifikationer
Kemiskt namn eller material (±)-Praeruptorin A
Kvantitet 100 mg
InChI formel InChI=1S/C21H22O7/c1-6-11(2)20(24)27-19-18(25-12(3)22)16-14(28-21(19,4)5)9-7-13-8-10-15(23)26-17(13)16/h6-10,18-19H,1-5H3/b11-6-/t18-,19-/m0/s1
Formel vikt 386.1366