Läs mer
Pralidoxime Methyl Sulphate, LoGiCal
Produktkod. 30458872
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30458872 10 mg
1 options
Produktkod. 30458872 Leverantör LoGiCal Leverantörsnummer LGCFOR0689.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pralidoxime Methyl Sulphate
CAS 1200-55-1
Analyt- eller komponentnamn Pralidoxime Methyl Sulphate
Frakt skick Ice pack (-20°C)
Molekylformel C7 H9 N2 O . C H3 O4 S
Certifikat och efterlevnad ISO 9001
Synonym Pralidoxime Methyl Sulphate
InChI formel InChI=1S/C7H8N2O.CH4O4S/c1-9-5-3-2-4-7(9)6-8-10;1-5-6(2,3)4/h2-6H,1H3;1H3,(H,2,3,4)
Lösningstyp Neat
LEDER COS(=O)(=O)[O-].C[n+]1ccccc1\C=N\O
IUPAC-namn (NE)-N-[(1-methylpyridin-1-ium-2-yl)methylidene]hydroxylamine;methyl sulfate
Industry Type Clinical Testing
Molekylvikt (g/mol) 248.26
Kvantitet 10 mg
Formel vikt 248.0467 g/mol