Läs mer
Pralmorelin Diacetate Salt, TRC
Produktkod. 30411509
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30411509 10 mg
1 options
Produktkod. 30411509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP70060510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pralmorelin Diacetate Salt
CAS 158861-67-7
Molekylformel C45H55N9O6 • 2C2H4O2
Kvantitet 10 mg
Synonym D-Alanyl-3-(2-naphthalenyl)-D-alanyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide
InChI formel InChI=1S/C45H55N9O6/c1-27(47)41(56)52-38(24-30-19-20-31-14-6-7-15-32(31)22-30)43(58)50-28(2)42(57)53-39(25-33-26-49-35-17-9-8-16-34(33)35)45(60)54-37(23-29-12-4-3-5-13-29)44(59)51-36(40(48)55)18-10-11-21-46/h3-9,12-17,19-20,22,26-28,36-39,49H,10-11,18,21,23-25,46-47H2,1-2H3,(H2,48,55)(H,50,58)(H,51,59)(H,52,56)(H,53,57)(H,54,60)/t27-,28+,36+,37-,38-,39+/m1/s1
LEDER O=C(N[C@H](C(N)=O)CCCCN)[C@H](NC([C@H](CC1=CNC2=C1C=CC=C2)NC([C@H](C)NC([C@H](NC([C@@H](C)N)=O)CC3=CC(C=CC=C4)=C4C=C3)=O)=O)=O)CC5=CC=CC=C5
Molekylvikt (g/mol) 817.9726005