Beskrivning
PRAM1 Polyclonal specifically detects PRAM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
Specifikationer
| Antigen | PRAM1 |
| Användningsområden | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassificering | Polyclonal |
| Konjugera | DyLight 594 |
| Utspädning | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulering | 50mM Sodium Borate |
| Gene Alias | MGC39864, PML-RAR, PML-RARA regulated adaptor molecule 1, PML-RARA target gene encoding an Adaptor Molecule-1, PML-RARA-regulated adapter molecule 1, PRAM, PRAM-1 |
| Värd art | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PRAM1 fragment. |
| Reningsmetod | Antigen Affinity-purified |
| Visa mer |