Learn More
PRAME Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11438-1-AP-20UL
 View more versions of this product
Product Code. 16888513
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
PRAME is a 509 amino acid protein belonging to the PRAME family consisting of 7 LRR (leucine-rich) repeats. It is a tumor antigen expressed in human melanomas and is recognized by cytolytic T lymphocytes. It′s overexpression frequently observed in human cancers confers growth or survival advantages by antagonizing retinoic acid receptor (RAR) signaling. PRAME serves as a candidate marker for monitoring childhood acute leukemia. It is expressed in testis and acute leukemias.
Specifications
Specifications
PRAME
Polyclonal
Unconjugated
PRAME
MAPE, OIP 4, OIP4, Opa interacting protein 4, PRAME
Rabbit
Antigen Affinity Chromatography
RUO
23532
-20°C
Liquid
Western Blot, Immunoprecipitation
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78395
PRAME
PRAME Fusion Protein Ag1906
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRAME Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated