missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Pranoprofen, MedChemExpress
Beskrivning
Pranoprofen is a non-steroidal anti-inflammatory agent (NSAID) for the research of keratitis or other ophthalmology diseases. Pranoprofen inhibit COX-1 and COX-2 enzymes, thus blocking arachidonic acid converted to eicosanoids and reducing prostaglandins synthesis.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pranoprofen |
| CAS | 52549-17-4 |
| Färg | Off-White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C15H13NO3 |
| Kvantitet | 500 mg |
| Löslighetsinformation | DMSO : ≥ 100 mg/mL (391.74 mM) |
| LEDER | CC(C(O)=O)C1=CC2=C(C=C1)OC3=NC=CC=C3C2 |
| Molekylvikt (g/mol) | 255.27 |
| Formel vikt | 255.27 |
| Visa mer |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?