missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Pravastatin Isopropyl Ester, Mikromol™
Description
Pravastatin Isopropyl Ester
Specifications
Specifications
| Kemiskt namn eller material | Pravastatin Isopropyl Ester |
| CAS | 2732719-88-7 |
| Analyt- eller komponentnamn | Pravastatin Isopropyl Ester |
| Frakt skick | Room Temperature |
| Molekylformel | C26 H42 O7 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Pravastatin Isopropyl Ester |
| InChI formel | InChI=1S/C26H42O7/c1-6-16(4)26(31)33-23-13-20(28)11-18-8-7-17(5)22(25(18)23)10-9-19(27)12-21(29)14-24(30)32-15(2)3/h7-8,11,15-17,19-23,25,27-29H,6,9-10,12-14H2,1-5H3/t16-,17-,19+,20+,21+,22-,23-,25-/m0/s1 |
| LEDER | CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)OC(C)C)[C@@H]12 |
| IUPAC-namn | propan-2-yl (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?