missing translation for 'onlineSavingsMsg'
Learn More

PRC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10110-2-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826283

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.
Specifications

Specifications

PRC1
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43663, Q99K43
ASE1, PRC1
Rabbit
IgG
20 μL
-20°C
Primary
233406, 308761, 9055
Western Blot
Unconjugated
PRC1
Liquid
PRC1
PRC1 Fusion Protein Ag0163
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers