missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PRC1 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826283
|
Proteintech
10110-2-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816283
|
Proteintech
10110-2-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.Specifications
PRC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43663, Q99K43 | |
233406, 308761, 9055 | |
PRC1 Fusion Protein Ag0163 | |
Primary | |
-20°C | |
PRC1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASE1, PRC1 | |
PRC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |