missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ PRDM12 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16687734
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16687734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16687734 Leverantör Invitrogen™ Leverantörsnummer PA5101259

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRDM12.

This gene encodes a transcriptional regulator of sensory neuronal specification that plays a critical role in pain perception. The encoded protein contains an N-terminal PRDI-BF1 and RIZ homology (PR) domain, a SET domain, and three C-terminal C2H2 zinc finger DNA-binding domains. Naturally occurring mutations in this gene are associated with congenital insensitivity to pain (CIP), and hereditary sensory and autonomic neuropathies (HSAN's) affecting peripheral sensory and autonomic neurons. Deregulation of this gene is associated with solid cancers and hematological malignancies including chronic myeloid leukemia.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen PRDM12
Användningsområden Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen PRDM12
Genaccessionsnr. A2AJ77, Q9H4Q4
Gene Alias Gm998; HSAN8; PFM9; PR domain 12; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PRDM12; PR-domain containing protein 12; PR-domain zinc finger protein 12; RP11-57C19.7
Gensymboler PRDM12
Värd art Rabbit
Immunogen A synthesized peptide derived from human PRDM12(Accession Q9H4Q4), corresponding to amino acid residues L198-C248.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 381359, 59335
Målarter Human, Mouse
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.