Läs mer
Saint Gobain Life Sciences Pre-drilled Rubber Stopper Without Holes
Produktkod. 10471155
Färg:
Blått
Förpackningsstorlek:
Paket med 100
Produktkod. 10471155
Brand: Saint Gobain Life Sciences 203006100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Use these stoppers for a variety of applications. Saint Gobain Life Sciences Rubber Stoppers Without Holes are ideal for chemical transfer.
  • Designed to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifikationer
Höjd (metrisk) 15mm
Kvantitet 100/ck.
Färg Blått
Diameter (metrisk) Botten 3.5 mm
Diameter (metrisk) Topp 6.5 mm
Material Natural Rubber
Produkttyp Rubber Stopper