missing translation for 'onlineSavingsMsg'
Läs mer

Merck Precoated TLC and PLC Glass Plates

Produktkod. 32237437
Change view
Klicka för att se tillgängliga alternativ
Sorbent:
Cellulose
Cellulose F
Kieselguhr F254
PEI Cellulose F
Silica Gel 60
Silica Gel 60 F254
Silica Gel 60 F254 AMD, extra thin
Silica Gel 60 F254 with preconcentrating zone
Silica Gel 60 RP-18 F254
Silica Gel 60 RP-18 F254 s
Silica Gel 60 RP-2 (silanized)
Silica Gel 60 RP-2 F254 (silanized)
Silica Gel 60 RP-8 F254
Silica Gel 60 RP-8 F254 s
Silica Gel 60 W F254 s
Silica Gel 60 with preconcentrating zone
Partikelstorlek:
10 to 12 μm
15 μm
5 to 20 μm
5 to 6 μm
Mått (L x B):
10 x 20 cm
2.5 x 7.5 cm
20 x 10 cm
20 x 20 cm
5 x 10 cm
5 x 20 cm
Kvantitet:
100/Pk.
12/Pk.
20/Pk.
25/Pk.
50/Pk.
Förpackningsstorlek:
Paket med 100
Paket med 12
Paket med 20
Paket med 25
Paket med 50
Varje
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Sorbent Partikelstorlek Mått (L x B) Kvantitet Förpackningsstorlek
32237437 Silica Gel 60 RP-2 (silanized) 10 to 12 μm 20 x 20 cm 25/Pk. -
15884660 Silica Gel 60 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
15825610 Silica Gel 60 F254 with preconcentrating zone 5 to 20 μm 20 x 20 cm 25/Pk. Paket med 25
15865810 Silica Gel 60 F254 10 to 12 μm 2.5 x 7.5 cm 100/Pk. Paket med 100
15805840 Silica Gel 60 RP-18 F254 s 5 to 20 μm 5 x 20 cm 50/Pk. Paket med 50
15885610 Silica Gel 60 F254 with preconcentrating zone 10 to 12 μm 10 x 20 cm 50/Pk. Paket med 50
15844680 Silica Gel 60 5 to 20 μm 20 x 20 cm 12/Pk. Paket med 12
15815610 Silica Gel 60 F254 AMD, extra thin 5 to 6 μm 20 x 10 cm 25/Pk. Varje
15885860 Silica Gel 60 W F254 s 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
15804670 PEI Cellulose F 5 to 20 μm 20 x 20 cm 25/Pk. Paket med 25
15864660 Cellulose F 15 μm 20 x 20 cm 25/Pk. Paket med 25
15875820 Silica Gel 60 RP-8 F254 10 to 12 μm 10 x 20 cm 50/Pk. Paket med 50
15855810 Silica Gel 60 10 to 12 μm 2.5 x 7.5 cm 100/Pk. Paket med 100
15894670 Kieselguhr F254 10 to 12 μm 20 x 20 cm 25/Pk. Varje
15865820 Silica Gel 60 RP-18 F254 s 10 to 12 μm 10 x 20 cm 50/Pk. Paket med 50
15815840 Silica Gel 60 RP-8 F254 s 10 to 12 μm 5 x 10 cm 25/Pk. Paket med 25
15815700 Silica Gel 60 15 μm 20 x 20 cm 20/Pk. Paket med 20
15824670 Cellulose F 15 μm 10 x 20 cm 50/Pk. Varje
15894660 Silica Gel 60 5 to 20 μm 5 x 20 cm 100/Pk. Varje
15875610 Silica Gel 60 with preconcentrating zone 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
15854680 Silica Gel 60 RP-2 F254 (silanized) 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
15845820 Silica Gel 60 RP-8 F254 s 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
15844660 Cellulose 15 μm 20 x 20 cm 25/Pk. Paket med 25
15865610 Silica Gel 60 with preconcentrating zone 15 μm 10 x 20 cm 50/Pk. Paket med 50
15814630 Silica Gel 60 10 to 12 μm 10 x 20 cm 50/Pk. Paket med 50
15844670 Cellulose 15 μm 10 x 20 cm 50/Pk. Varje
15825840 Silica Gel 60 RP-18 F254 10 to 12 μm 20 x 20 cm 25/Pk. Paket med 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 32237437 Leverantör Merck Leverantörsnummer 1057460001
 Begränsad artikel
Beräknad leverans: 04-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Average particle size is 15μm with a 5-20μm distribution

  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60 Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates

Certifikat

ISO 9001:2000

†With acid-stable fluorescent indicator.

‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.

TRUSTED_SUSTAINABILITY

Specifikationer

Stödmaterial Glass
Certifikat och efterlevnad ISO 9001:2000
Material Silica 60
Partikelstorlek 10 to 12 μm
Porstorlek 60 Å
Kvantitet 25/Pk.
Typ RP-modified Silica TLC Plate
För användning med (applikation) Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems
Sorbent Silica Gel 60 RP-2 (silanized)
Tjocklek (metrisk) 250 μm
Mått (L x B) 20 x 20 cm
Längd (metrisk) 20 cm
Bredd (metrisk) 20 cm
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.