Läs mer
Prednisolone 21-Propionate, TRC
Produktkod. 30136144
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136144 5 g
18063720 1 g
18072030 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136144 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7039005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5740-62-5
Molekylformel C24 H32 O6
Molekylvikt (g/mol) 416.51
Synonym Pregna-1,4-diene-3,20-dione, 11,17-dihydroxy-21-(1-oxopropoxy)-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-, 21-propionate (6CI,7CI,8CI), (11β)-11,17-Dihydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Prednisolone 21-propionate, 11β,17-Dihydroxy-3,20-dioxopregna-1,4-dien-21-yl Propanoate, Prednisolone 21-Propanoate, Prednicarbate Imp. C (EP)
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material prednisolone-21-propionate
Kvantitet 5 g
InChI formel InChI=1S/C24H32O6/c1-4-20(28)30-13-19(27)24(29)10-8-17-16-6-5-14-11-15(25)7-9-22(14,2)21(16)18(26)12-23(17,24)3/h7,9,11,16-18,21,26,29H,4-6,8,10,12-13H2,1-3H3/t16-,17-,18-,21+,22-,23-,24-/m0/s1
Formel vikt 416.2199