Läs mer
Prednisolone 21-Sulfate Sodium Salt, TRC
Produktkod. 30134087
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134087 100 mg
30010759 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134087 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP703755100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3664-95-7
Molekylformel C21H27NaO8S
Molekylvikt (g/mol) 462.49
Synonym (11β)-11,17-Dihydroxy-21-(sulfooxy)-pregna-1,4-diene-3,20-dione Sodium Salt, 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(Hydrogen Sulfate) Monosodium Salt;
IUPAC-namn sodium;[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] sulfate
LEDER Cl.COc1c(cc(cc1)C(=O)C(C)N)S(=O)(N)=O
Specifikationer
Kemiskt namn eller material Prednisolone 21-Sulfate Sodium Salt
Kvantitet 100 mg
InChI formel InChI=1S/C21H28O8S.Na/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;/h5,7,9,14-16,18,23,25H,3-4,6,8,10-11H2,1-2H3,(H,26,27,28);/q;+1/p-1/t14-,15-,16-,18+,19-,20-,21-;/m0./s1
Formel vikt 462.1324