Läs mer
Prednisolone Caproate, TRC
Produktkod. 30133149
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133149 1 g
30004002 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133149 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7037701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69164-69-8
Molekylformel C27 H38 O6
Molekylvikt (g/mol) 458.59
Synonym (11β)-11,17-Dihydroxy-21-[(1-oxohexyl)oxy]-pregna-1,4-diene-3,20-dione, 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-Hexanoate
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] hexanoate
LEDER CCCCCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Prednisolone hexanoate
Kvantitet 1 g
InChI formel InChI=1S/C27H38O6/c1-4-5-6-7-23(31)33-16-22(30)27(32)13-11-20-19-9-8-17-14-18(28)10-12-25(17,2)24(19)21(29)15-26(20,27)3/h10,12,14,19-21,24,29,32H,4-9,11,13,15-16H2,1-3H3/t19-,20-,21-,24+,25-,26-,27-/m0/s1
Formel vikt 458.2668