Läs mer
Prednisolone Hemisuccinate, TRC
Produktkod. 30134548
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134548 10 g
18070920 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134548 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP70376010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2920-86-7
Molekylformel C25 H32 O8
Molekylvikt (g/mol) 460.52
Synonym (11beta)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(hydrogen succinate), Prednisolone 21-O-beta-carbonylpropionic acid, Prednisolone 21-hemisuccinate, Prednisolone 21-succinate, Prednisolone bisuccinate, Prednisolone hemisuccinate, Prednisolone succinate, Prednisolut, delta1-Hydrocortisone 21-hemisuccinate
IUPAC-namn 4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)CCC(=O)O
Specifikationer
Kemiskt namn eller material Prednisolone hemisuccinate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C25H32O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h7,9,11,16-18,22,27,32H,3-6,8,10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1
Formel vikt 460.2097
Procent renhet >95%