Läs mer
Prednisolone Pivalate, LoGiCal
Produktkod. 30461630
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461630 10 mg
1 options
Produktkod. 30461630 Leverantör LoGiCal Leverantörsnummer LGCFOR0304.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Prednisolone pivalate
CAS 1107-99-9
Analyt- eller komponentnamn Prednisolone pivalate
Frakt skick Room Temperature
Molekylformel C26 H36 O6
Certifikat och efterlevnad ISO 9001
Synonym Prednisolone Pivalate, Mecortolon, PTMA, Prednisolone 21-trimethylacetate, Prednisolone pivalate, Prednisolone trimethylacetate, Ultracortenol, Ultracorterenol, Vecortenol, (11beta)-21-(2,2-Dimethyl-1-oxopropoxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-pivalate, 11beta,17alpha-Dihydroxy-21-pivaloyloxypregna-1,4-diene-3,20-dione
InChI formel InChI=1S/C26H36O6/c1-23(2,3)22(30)32-14-20(29)26(31)11-9-18-17-7-6-15-12-16(27)8-10-24(15,4)21(17)19(28)13-25(18,26)5/h8,10,12,17-19,21,28,31H,6-7,9,11,13-14H2,1-5H3/t17-,18-,19-,21+,24-,25-,26-/m0/s1
Lösningstyp Neat
LEDER CC(C)(C)C(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanoate
Industry Type Clinical Testing
Molekylvikt (g/mol) 444.56
Kvantitet 10 mg
Formel vikt 444.2512 g/mol