Läs mer
Prednisolone Pivalate, TRC
Produktkod. 30416068
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416068 100 mg
30410817 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416068 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP703880100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1107-99-9
Molekylformel C26 H36 O6
Molekylvikt (g/mol) 444.56
Synonym Prednisolone Pivalate, (11β)-21-(2,2-Dimethyl-1-oxopropoxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-pivalate, 11β,17α-Dihydroxy-21-pivaloyloxypregna-1,4-diene-3,20-dione, Mecortolon, PTMA, Prednisolone 21-trimethylacetate, Prednisolone pivalate, Prednisolone trimethylacetate, Ultracortenol, Ultracorterenol, Vecortenol
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2-dimethylpropanoate
LEDER CC(C)(C)C(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Prednisolone pivalate
Kvantitet 100 mg
InChI formel InChI=1S/C26H36O6/c1-23(2,3)22(30)32-14-20(29)26(31)11-9-18-17-7-6-15-12-16(27)8-10-24(15,4)21(17)19(28)13-25(18,26)5/h8,10,12,17-19,21,28,31H,6-7,9,11,13-14H2,1-5H3/t17-,18-,19-,21+,24-,25-,26-/m0/s1
Formel vikt 444.2512