Läs mer
Prednisolone Sodium Metasulfobenzoate, Mikromol™
Produktkod. 17951636
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
17951636 250 mg
1 options
Produktkod. 17951636 Leverantör Mikromol Leverantörsnummer MM0484.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Prednisolone Sodium Metasulfobenzoate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Prednisolone Sodium Metasulfobenzoate
CAS 630-67-1
Analyt- eller komponentnamn Prednisolone Sodium Metasulfobenzoate
Frakt skick Room Temperature
Molekylformel C28 H31 O9 S . Na
Certifikat och efterlevnad ISO 17034
Synonym ATL 2502, Cortico-Sol, Predenema, Predfoam, Prednisolone 21-m-sulfobenzoate sodium, Prednisolone 21-m-sulfobenzoate sodium salt, Prednisolone metasulfobenzoate sodium, Prednisolone sodium metasulfobenzoate, Prednisolone sodium metazoate, Predocol, Pregna-1,4-diene-3,20-dione, 11,17-dihydroxy-21-[(3-sulfobenzoyl)oxy]-, sodium salt (1:1), (11beta)-, Pregna-1,4-diene-3,20-dione, 11,17-dihydroxy-21-[(3-sulfobenzoyl)oxy]-, monosodium salt, (11beta)- (9CI), Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-, 21-(m-sulfobenzoate), monosodium salt (8CI), Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-, 21-m-sulfobenzoate, sodium salt (7CI), Benzoic acid, m-sulfo-, 21-ester with 11beta,17,21-trihydroxypregna-1,4-diene-3,20-dione, monosodium salt (8CI)
InChI formel InChI=1S/C28H32O9S.Na/c1-26-10-8-18(29)13-17(26)6-7-20-21-9-11-28(33,27(21,2)14-22(30)24(20)26)23(31)15-37-25(32)16-4-3-5-19(12-16)38(34,35)36;/h3-5,8,10,12-13,20-22,24,30,33H,6-7,9,11,14-15H2,1-2H3,(H,34,35,36);/q;+1/p-1/t20-,21-,22-,24+,26-,27-,28-;/m0./s1
LEDER [Na+].C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)c5cccc(c5)S(=O)(=O)[O-]
IUPAC-namn sodium;3-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]carbonylbenzenesulfonate
Molekylvikt (g/mol) 566.6
Kvantitet 250 mg
Formel vikt 566.1586 g/mol