missing translation for 'onlineSavingsMsg'
Learn More

Prednisone 21-Acetate, TRC

Product Code. 30425381
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30425381 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425381 Supplier Toronto Research Chemicals Supplier No. TRCP703825250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Prednisone Acetate
CAS 125-10-0
Molekylformel C23 H28 O6
Kvantitet 250 mg
Synonym Prednisone Acetate, 21-(Acetyloxy)-17-hydroxypregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione 21-acetate, 21-Acetoxy-17α-hydroxypregna-1,4-diene-3,11,20-trione, Cortancyl, Delcortin, Delta-Corlin, Deltalone, Ferrosan, NSC 10965, Nisone, Prednisone 21-acetate, Prednisone acetate, Δ1-Cortisone 21-acetate, Δ1-Dehydrocortisone acetate
InChI formel InChI=1S/C23H28O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-17,20,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC-namn [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 400.46
Formel vikt 400.1886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.