Läs mer
Prednisone 21-Acetate, TRC
Produktkod. 30418163
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30418163 100 mg
1 options
Produktkod. 30418163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP703825100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Prednisone Acetate
CAS 125-10-0
Molekylformel C23 H28 O6
Kvantitet 100 mg
Synonym Prednisone Acetate, 21-(Acetyloxy)-17-hydroxypregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione 21-acetate, 21-Acetoxy-17α-hydroxypregna-1,4-diene-3,11,20-trione, Cortancyl, Delcortin, Delta-Corlin, Deltalone, Ferrosan, NSC 10965, Nisone, Prednisone 21-acetate, Prednisone acetate, Δ1-Cortisone 21-acetate, Δ1-Dehydrocortisone acetate
InChI formel InChI=1S/C23H28O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-17,20,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC-namn [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 400.46
Formel vikt 400.1886