Learn More
Honeywell™ Prefilter Pads
Prefilter Pads are used in conjunction with a retaining clip or adapter on Honeywell™ Twin-Filter Air-Purifying Respirators
48.00 SEK - 1070.00 SEK
Specifications
Certifications/Compliance | PPE Cat. 3; ISO 9001/2000 |
---|---|
Certifications (EN) | EN 143 |
Product Type | Prefilter Pad |
Product Code | Brand | Cartridge Type | Design Type | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cartridge Type | Design Type | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | ||
12682743
|
Honeywell™
1002801 |
Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators | Requires Retaining Cover (1001590 or 1001606) | Respiratory Protection (P2, Dust/Fume) | Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators |
48.00 SEK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
15806645
|
Honeywell™
N7500P3O |
Bayonet Gas/Vapor Cartridges on N-Series Respirators | Requires Adapter (N06575035) | Respiratory Protection (P2/P3, Particulates/Odor) | Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators |
1070.00 SEK
1 pair |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
15896635
|
Honeywell™
N7500P3 |
Bayonet Gas/Vapor Cartridges on N-Series Respirators | Requires Adapter (N06575035) | Respiratory Protection (P2/P3, Particulates) | Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators |
138.00 SEK
1 pair |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
11847171
|
Honeywell™
1002800 |
Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators | Requires Retaining Cover (1001590 or 1001606) | Respiratory Protection (P1, Dust) | Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Product Type: Prefilter Pads
- For Use With: Honeywell™ Twin-Filter Air-Purifying Respirators
Specifications
PPE Cat. 3; ISO 9001/2000 | |
EN 143 | |
Prefilter Pad |