Läs mer
uvex Preformed Mask silv-Air p 7100 FFP1
Produktkod. 17562945 Handla allt Uvex Produkter
Change view
Filter Type: FFP1
Produktkod.
17562945 Filter Type: FFP1
1 options
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
Produktkod. 17562945 Leverantör uvex Leverantörsnummer 8707100
  • ONLINE EXCLUSIVE PRICE
    461.00 SEK
    435.27 SEK / Paket med 20
    Save 25.73 SEK
    5% Av
I lager
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering preformed mask
  • Metal-free processing
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
  • Color: silver-gray
Protection Features
  • FFP1 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
TRUSTED_SUSTAINABILITY
Specifikationer
Certifieringar/efterlevnad (Europa) EN 149 (D marked)
Färg White
Designtyp Cup/Molded
Disponibel Single-use
Filtertyp Particulate
Material Polypropylene, Polyester
Produkttyp Particulate Respirator
Storlek Standard
Med utandningsventil No
Filtereffektivitet FFP1
Sterilitet Non-sterile