Läs mer
Pregabalin, Lipomed
Produktkod. 30460450
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30460450 100 mg
1 options
Produktkod. 30460450 Leverantör Lipomed Leverantörsnummer LPMPRE1475100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Pregabalin
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pregabalin
CAS 148553-50-8
Analyt- eller komponentnamn Pregabalin
Frakt skick Room Temperature
Molekylformel C8 H17 N O2
Synonym Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3 S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (S)-, (2 S)-3-(Aminomethyl)-5-methylhexanoic acid, (3 S)-3-(Aminomethyl)-5-methylhexanoic acid, 3 S-(aminomethyl)-5-methyl-hexanoic acid, (S)-(+)-3-(Aminomethyl)-5-methylhexanoic acid, (S)-3-(Ammoniomethyl)-5-methylhexanoate, (S)-Pregabalin, CI 1008, Gabanext 75, Lyrica, Mahagaba-75, Maxgalin 75, Neugaba 75, PD 144723, Pregabalin, Pregalin 75, (S)-Isobutylgaba
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER CC(C)C[C@H](CN)CC(=O)O
IUPAC-namn (3 S)-3-(aminomethyl)-5-methylhexanoic acid
Industry Type Pharmaceutical
Molekylvikt (g/mol) 159.23
Kvantitet 100 mg
Formel vikt 159.1259 g/mol