missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Pregabalin Methyl Ester Hydrochloride, Mikromol™
Beskrivning
Pregabalin Methyl Ester Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pregabalin Methyl Ester Hydrochloride |
| CAS | 714230-22-5 |
| Analyt- eller komponentnamn | Pregabalin Methyl Ester Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C9 H19 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | (S)-Pregabalin Methyl Ester Hydrochloride, Hexanoic acid, 3-(aminomethyl)-5-methyl-, methyl ester, hydrochloride (1:1), (3S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, methyl ester, hydrochloride, (3S)- (9CI) |
| InChI formel | InChI=1S/C9H19NO2.ClH/c1-7(2)4-8(6-10)5-9(11)12-3;/h7-8H,4-6,10H2,1-3H3;1H/t8-;/m0./s1 |
| LEDER | Cl.COC(=O)C[C@@H](CN)CC(C)C |
| IUPAC-namn | methyl (3S)-3-(aminomethyl)-5-methylhexanoate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?