Läs mer
Pregna-4,9(11)-diene-3,20-dione (90%), TRC
Produktkod. 30427556
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427556 50 mg
30410905 100 mg
30427275 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427556 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP70497050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17652-16-3
Molekylformel C21 H28 O2
Molekylvikt (g/mol) 312.45
Synonym Pregna-4,9(11)-diene-3,20-dione, 9-Dehydroprogesterone, δ9(11)-Progesterone, Progesterone Imp. K (EP)
IUPAC-namn (8S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-1,2,6,7,8,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)C3=CC[C@]12C
Specifikationer
Kemiskt namn eller material Pregna-4,9(11)-diene-3,20-dione
Kvantitet 50 mg
InChI formel InChI=1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h9,12,16-18H,4-8,10-11H2,1-3H3/t16-,17+,18-,20-,21+/m0/s1
Formel vikt 312.2089