Läs mer
Pregnanolone Sulfate Pyridinium Salt, TRC
Produktkod. 30423660
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423660 5 mg
30418453 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423660 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7113205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124107-39-7
Molekylformel C26H39NO5S
Molekylvikt (g/mol) 477.66
Synonym 3α-Hydroxy-5β-pregnan-20-one Sulfate Pyridinium Salt, 5β-Pregnan-3α-ol-20-one Sulfate Pyridinium Salt, 5β-Pregnane-3α-yl-20-one Sulfate Pyridinium Salt
LEDER CC(=O)[C@H]1CC[C@@H]2[C@@]1(CC[C@H]3[C@H]2CC[C@H]4[C@@]3(CC[C@H](C4)OS(=O)(=O)[O-])C)C.C1=CC=[NH+]C=C1
Specifikationer
Kemiskt namn eller material Pregnanolone Sulfate Pyridinium Salt
Kvantitet 5 mg
InChI formel InChI=1S/C21H34O5S.C5H5N/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;1-2-4-6-5-3-1/h14-19H,4-12H2,1-3H3,(H,23,24,25);1-5H/t14-,15-,16+,17-,18+,19+,20+,21-;/m1./s1
Formel vikt 477.2549