Läs mer
Pregnenolone-3-sulfate Sodium Salt, TRC
Produktkod. 30136081
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136081 250 mg
30007928 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP712220250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1852-38-6
Molekylformel C21H31NaO5S
Molekylvikt (g/mol) 418.52
Synonym 3-(Sulfooxy)pregn-5-en-20-one Sodium Salt, (3beta-3-(Sulfooxy)pregn-5-en-20-one Sodium Salt, 3beta-Hydroxypregn-5-en-20-one Hydrogen Sulfate Sodium Salt, Pregnenolone Sulfate Sodium Salt;
IUPAC-namn sodium;[(3S,8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] sulfate
LEDER [Na+].CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Pregnenolone-3-sulfate Sodium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C21H32O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,15-19H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t15-,16-,17+,18-,19-,20-,21+;/m0./s1
Formel vikt 418.179