Läs mer
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Produktkod. 12313850
Change view
Slangstorlek:
3/16 x 1/16 tum
5/32 x 1/16 tum
Förpackningsstorlek:
Paket med 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Slangstorlek Förpackningsstorlek
12313850 5/32 x 1/16 tum Paket med 10
12323850 3/16 x 1/16 tum Paket med 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
Produktkod. 12313850 Leverantör Ark-Plas Products™ Leverantörsnummer MAD80DF0
  • ONLINE EXCLUSIVE PRICE
    179.00 SEK
    7.60 SEK / Paket med 10
    Save 171.40 SEK
    95% Av
I lager
\r\n Utg\u00E5ngen artikel:\r\n
\r\n Den h\u00E4r artikeln utg\u00E5r ur v\u00E5rt sortiment n\u00E4r v\u00E5rt lager \u00E4r slut.\r\n
\r\n
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
  • Made of Kynar
  • Economical straight barbed
  • Easily connect fluid paths without the use of tools
Specifikationer
Material Fluororesin
Slangstorlek 5/32 x 1/16 tum
Färg Natural
Produkttyp Straight reducing connector