Läs mer
Previtamin D3 (>80%), TRC
Produktkod. 18053750
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18053750 1 mg
17909969 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18053750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7135101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1173-13-3
Molekylformel C27 H44 O
Molekylvikt (g/mol) 384.64
Synonym (1S)-3-[(1Z)-2-[(1R,3aR,7aR)-1-[(1R)-1,5-Dimethylhexyl]-2,3,3a,6,7,7a-hexahydro-7a-methyl-1H-inden-4-yl]ethenyl]-4-methyl-3-cyclohexen-1-ol, 9,10-Secocholesta-5(10),6,8-trien-3-ol, (3β,6Z)- (9CI), 9,10-Secocholesta-5(10),6,8-trien-3β-ol (8CI), Precalciferol (6CI), Previtamin D3 (7CI), Precalciferol3, Precholecalciferol, Previtamin D, Prächolecalciferol
IUPAC-namn (1S)-3-[(Z)-2-[(1R,3aR,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-1,2,3,3a,6,7-hexahydroinden-4-yl]ethenyl]-4-methylcyclohex-3-en-1-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C(=CCC[C@]12C)\C=C/C3=C(C)CC[C@H](O)C3
Specifikationer
Kemiskt namn eller material Previtamin D3
Kvantitet 1 mg
InChI formel InChI=1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h10,12-13,19,21,24-26,28H,6-9,11,14-18H2,1-5H3/b13-12-/t21-,24+,25-,26+,27-/m1/s1
Formel vikt 384.3392