Läs mer
Prifinium Bromide, TRC
Produktkod. 30132951
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132951 1 g
30137120 250 mg
30011938 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7175001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4630-95-9
Molekylformel C22 H28 N . Br
Molekylvikt (g/mol) 386.37
Synonym Pyrrolidinium, 3-(diphenylmethylene)-1,1-diethyl-2-methyl-, bromide (1:1), 3-(Diphenylmethylene)-1,1-diethyl-2-methylpyrrolidinium bromide (7CI), Pyrrolidinium, 3-(diphenylmethylene)-1,1-diethyl-2-methyl-, bromide (8CI,9CI), 3-(Diphenylmethylene)-1-ethyl-2-methylpyrrolidine ethyl bromide, Padrin, Prifinium bromide, Pyrodifenium bromide, Riabal
IUPAC-namn 3-benzhydrylidene-1,1-diethyl-2-methylpyrrolidin-1-ium;bromide
LEDER [Br-].CC[N+]1(CC)CCC(=C(c2ccccc2)c3ccccc3)C1C
Specifikationer
Kemiskt namn eller material Prifinium Bromide
Kvantitet 1 g
InChI formel InChI=1S/C22H28N.BrH/c1-4-23(5-2)17-16-21(18(23)3)22(19-12-8-6-9-13-19)20-14-10-7-11-15-20;/h6-15,18H,4-5,16-17H2,1-3H3;1H/q+1;/p-1
Formel vikt 385.1405