Läs mer
Prilocaine EP Impurity A, Mikromol™
Produktkod. 30051868
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051868 100 mg
30052168 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051868 Leverantör Mikromol Leverantörsnummer MM0563.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Prilocaine EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (RS)-2-Chloro-N-(2-methylphenyl)propanamide
CAS 19281-31-3
Analyt- eller komponentnamn (RS)-2-Chloro-N-(2-methylphenyl)propanamide
Frakt skick Room Temperature
Molekylformel C10 H12 Cl N O
Certifikat och efterlevnad ISO 17025
Synonym Prilocaine Hydrochloride Imp. A (EP), Prilocaine Imp. A (EP), (RS)-2-Chloro-N-(2-methylphenyl)propanamide, Prilocaine Hydrochloride Impurity A, Prilocaine Impurity A
InChI formel InChI=1S/C10H12ClNO/c1-7-5-3-4-6-9(7)12-10(13)8(2)11/h3-6,8H,1-2H3,(H,12,13)
LEDER O=C(NC=1C=CC=CC1C)C(Cl)C
IUPAC-namn 2-chloro-N-(2-methylphenyl)propanamide
Molekylvikt (g/mol) 197.66
Kvantitet 100 mg
Formel vikt 197.0607 g/mol