Learn More
Invitrogen™ PRIMER 5'BDJ TR-X 200 NMOL
Custom Primer
Brand:  Invitrogen™ 11747011
Product Code. 10328333
  • 3555.00 SEK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' BDJ TR-X Primer, 200 nmol
5' BDJ TR-X Primer
5' Labeled
200 nmol