Läs mer
Primidone-d5, TRC
Produktkod. 17995115
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17995115 1 mg
17977359 10 mg
17959079 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17995115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP7340021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73738-06-4
Molekylformel C12 2H5 H9 N2 O2
Molekylvikt (g/mol) 223.28
Synonym 4,6(1H,5H)-Pyrimidinedione, 5-(ethyl-d5)dihydro-5-phenyl- (9CI), Primidone-d5, Primidone-D5 (ethyl-D5), 5-(Ethyl-d5)-5-phenyldihydropyrimidine-4,6(1H,5H)-dione, 2-Desoxyphenobarbital-d5
IUPAC-namn 5-(1,1,2,2,2-pentadeuterioethyl)-5-phenyl-1,3-diazinane-4,6-dione
LEDER [2H]C([2H])([2H])C([2H])([2H])C1(C(=O)NCNC1=O)c2ccccc2
Specifikationer
Kemiskt namn eller material Primidone-d5
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C12H14N2O2/c1-2-12(9-6-4-3-5-7-9)10(15)13-8-14-11(12)16/h3-7H,2,8H2,1H3,(H,13,15)(H,14,16)/i1D3,2D2
Formel vikt 223.1369
Procent renhet >95%